ENA405195482 Enamine

3-(oxolan-2-yl)propan-1-ol

  • MDL number MFCD01725793
  • Cas Number : 767-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85966
Availability details for
Quantity
  • Availability
  • Add to Cart