ENA405195488 Enamine

3-[(2-fluorophenyl)methyl]-1-(4-methylbenzenesulfonyl)azetidine

  • MDL number MFCD25970558
  • Cas Number : 1356739-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93305
Availability details for
Quantity
  • Availability
  • Add to Cart