ENA405195496 Enamine

4-amino-3-methyl-2-(trifluoromethyl)benzonitrile

  • MDL number MFCD09835408
  • Cas Number : 573764-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97264

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97264
Availability details for
Quantity
  • Availability
  • Add to Cart