ENA405195498 Enamine

4-bromo-2-chloronaphthalen-1-amine

  • MDL number MFCD21935644
  • Cas Number : 51670-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99167
Availability details for
Quantity
  • Availability
  • Add to Cart