ENA408615332 Enamine

methyl octahydropyrido[2,1-c]morpholine-3-carboxylate

  • MDL number MFCD26407530
  • Cas Number : 1461713-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121872
Availability details for
Quantity
  • Availability
  • Add to Cart