ENA408615333 Enamine

7-methoxy-3,4-dihydronaphthalene-2-sulfonamide

  • MDL number MFCD26407533
  • Cas Number : 1461715-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-123838

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123838
Availability details for
Quantity
  • Availability
  • Add to Cart