ENA408615338 Enamine

3-[1-(3-methoxyphenyl)-3,5-dimethyl-1H-pyrazol-4-yl]morpholine

  • MDL number MFCD26407552
  • Cas Number : 1461708-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129798
Availability details for
Quantity
  • Availability
  • Add to Cart