ENA408615340 Enamine

3-[5-(1-methyl-1H-pyrazol-4-yl)-1,2,4-oxadiazol-3-yl]morpholine

  • MDL number MFCD26275812
  • Cas Number : 1461705-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130284

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130284
Availability details for
Quantity
  • Availability
  • Add to Cart