ENA408615343 Enamine

2-(bromomethyl)-5-methyl-4H,7H-[1,2,4]triazolo[1,5-a]pyrimidin-7-one

  • MDL number MFCD26407567
  • Cas Number : 1461706-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231...233
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131040

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131040
Availability details for
Quantity
  • Availability
  • Add to Cart