ENA408615351 Enamine

5-chloro-7-nitro-2,3-dihydro-1-benzofuran

  • MDL number MFCD26407579
  • Cas Number : 150805-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131794

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131794
Availability details for
Quantity
  • Availability
  • Add to Cart