ENA408615354 Enamine

tert-butyl N-[1-(2-chlorophenyl)-2-oxocyclohexyl]-N-methylcarbamate

  • MDL number MFCD26407585
  • Cas Number : 1461707-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132319

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132319
Availability details for
Quantity
  • Availability
  • Add to Cart