ENA408615360 Enamine

1,3-diethyl 2-(4-fluoro-2-nitrobenzoyl)propanedioate

  • MDL number MFCD26407597
  • Cas Number : 108934-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74...76
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133066

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133066
Availability details for
Quantity
  • Availability
  • Add to Cart