ENA408615362 Enamine

cyclobutyl 6-chloro-2-cyclobutoxypyridine-3-carboxylate

  • MDL number MFCD26407599
  • Cas Number : 1461713-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133254
Availability details for
Quantity
  • Availability
  • Add to Cart