ENA408615363 Enamine

4-(2-methoxyethoxy)-3-nitrobenzonitrile

  • MDL number MFCD26407601
  • Cas Number : 1461713-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133341
Availability details for
Quantity
  • Availability
  • Add to Cart