ENA408615364 Enamine

5-(benzylsulfanyl)-3-ethyl-[1,2]oxazolo[5,4-b]pyridine

  • MDL number MFCD26407602
  • Cas Number : 1461708-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133651

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133651
Availability details for
Quantity
  • Availability
  • Add to Cart