ENA408615365 Enamine

1-[(1-benzyl-4-methylpiperidin-4-yl)methyl]-4-methylpiperazine

  • MDL number MFCD26407608
  • Cas Number : 1461714-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133896
Availability details for
Quantity
  • Availability
  • Add to Cart