ENA408615367 Enamine

6-bromo-7-(hydroxymethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD26407609
  • Cas Number : 1461713-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-134062

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134062
Availability details for
Quantity
  • Availability
  • Add to Cart