ENA408615368 Enamine

2-cyclopropyl-3-methyloxolan-3-amine

  • MDL number MFCD26407610
  • Cas Number : 1461707-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134123
Availability details for
Quantity
  • Availability
  • Add to Cart