ENA408615370 Enamine

4,5-dibromo-N-(2,2-diethoxyethyl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD26407616
  • Cas Number : 1461713-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134297

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134297
Availability details for
Quantity
  • Availability
  • Add to Cart