ENA408615374 Enamine

3-(4-methylbenzoyl)-5H,6H,7H-pyrrolo[1,2-a]imidazol-2-amine

  • MDL number MFCD26407622
  • Cas Number : 1377567-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134518

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134518
Availability details for
Quantity
  • Availability
  • Add to Cart