ENA408615375 Enamine

[6-(aminomethyl)piperidin-2-yl]methanol

  • MDL number MFCD26407623
  • Cas Number : 1461714-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134529
Availability details for
Quantity
  • Availability
  • Add to Cart