ENA408615377 Enamine

1-(azetidin-3-yl)-4-phenyl-1H-1,2,3-triazole hydrochloride

  • MDL number MFCD26407626
  • Cas Number : 1461713-51-8
  • Molecular Weight : 236.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(azetidin-3-yl)-4-phenyl-1H-1,2,3-triazole hydrochloride
Inchi Code 1S/C11H12N4.ClH/c1-2-4-9(5-3-1)11-8-15(14-13-11)10-6-12-7-10;/h1-5,8,10,12H,6-7H2;1H
Inchi Key BJHCRURCMQOPID-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-134701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134701
Availability details for
Quantity
  • Availability
  • Add to Cart