ENA408615378 Enamine

[4-(4-methylbenzenesulfonyl)phenyl]methanol

  • MDL number MFCD26407627
  • Cas Number : 1461713-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134881

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134881
Availability details for
Quantity
  • Availability
  • Add to Cart