ENA408615379 Enamine

ethyl 2-methyloxirane-2-carboxylate

  • MDL number MFCD26407631
  • Cas Number : 96685-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135026

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135026
Availability details for
Quantity
  • Availability
  • Add to Cart