ENA408615380 Enamine

(2S)-N-methyloxolane-2-carboxamide

  • MDL number MFCD26407634
  • Cas Number : 1222386-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135137
Availability details for
Quantity
  • Availability
  • Add to Cart