ENA408615383 Enamine

tert-butyl N-{bicyclo[2.2.1]hept-5-en-2-yl}carbamate

  • MDL number MFCD26407638
  • Cas Number : 1242273-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135171

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135171
Availability details for
Quantity
  • Availability
  • Add to Cart