ENA408615385 Enamine

N-(4-acetyl-2-fluorophenyl)methanesulfonamide

  • MDL number MFCD25967524
  • Cas Number : 824937-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135509

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135509
Availability details for
Quantity
  • Availability
  • Add to Cart