ENA408615391 Enamine

1-(2-methoxyethyl)-4,5,6,7-tetrahydro-1H-indazol-7-amine

  • MDL number MFCD26407655
  • Cas Number : 1461706-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135772
Availability details for
Quantity
  • Availability
  • Add to Cart