ENA408615392 Enamine

3-(azocan-1-yl)-1-(4-chlorophenyl)propan-1-one hydrochloride

  • MDL number MFCD26407656
  • Cas Number : 1222-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-135773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135773
Availability details for
Quantity
  • Availability
  • Add to Cart