ENA408615393 Enamine

1-(1,3-thiazol-2-yl)pyrrolidine-2-carboxylic acid hydrochloride

  • MDL number MFCD26407657

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218...220
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135779

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135779
Availability details for
Quantity
  • Availability
  • Add to Cart