ENA408615404 Enamine

4-[(3-cyclopropyl-1,2,4-oxadiazol-5-yl)methyl]aniline hydrochloride

  • MDL number MFCD26407675
  • Cas Number : 1461714-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136026

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136026
Availability details for
Quantity
  • Availability
  • Add to Cart