ENA408615406 Enamine

N-(2,6-dimethylphenyl)-3-hydroxynaphthalene-2-carboxamide

  • MDL number MFCD26407678
  • Cas Number : 92886-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136030
Availability details for
Quantity
  • Availability
  • Add to Cart