ENA408615408 Enamine

ethyl 2-cyano-2-(2-fluorophenyl)-2-methylacetate

  • MDL number MFCD26407680
  • Cas Number : 1461705-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136045

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136045
Availability details for
Quantity
  • Availability
  • Add to Cart