ENA408615410 Enamine

pent-1-yn-3-amine hydrochloride

  • MDL number MFCD26407682
  • Cas Number : 50329-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136061
Availability details for
Quantity
  • Availability
  • Add to Cart