ENA408615412 Enamine

1-(4-methylpyridin-2-yl)-5-propyl-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD26407689
  • Cas Number : 1461704-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136132
Availability details for
Quantity
  • Availability
  • Add to Cart