ENA408615416 Enamine

(1s,4s)-4-(4-methylpiperazin-1-yl)cyclohexan-1-amine trihydrochloride

  • MDL number MFCD26407704
  • Cas Number : 1461689-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-136225
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart