ENA408615417 Enamine

1-[(4-formylphenyl)sulfanyl]-N,N-dimethylformamide

  • MDL number MFCD26407705
  • Cas Number : 19654-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-136234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136234
Availability details for
Quantity
  • Availability
  • Add to Cart