ENA408615418 Enamine

1-{[4-(hydroxymethyl)phenyl]sulfanyl}-N,N-dimethylformamide

  • MDL number MFCD26407706
  • Cas Number : 1461704-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136235

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136235
Availability details for
Quantity
  • Availability
  • Add to Cart