ENA408615420 Enamine

5-({[(oxolan-2-yl)methyl]amino}methyl)furan-2-carboxylic acid hydrochloride

  • MDL number MFCD26407709
  • Cas Number : 1461709-31-8
  • Molecular Weight : 261.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-136259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136259
Availability details for
Quantity
  • Availability
  • Add to Cart