ENA408615436 Enamine

2-hydroxy-5-(2-{[(2-methoxyphenyl)methyl]amino}-1,3-thiazol-4-yl)benzamide hydrochloride

  • MDL number MFCD26407728
  • Cas Number : 1461708-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136403

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136403
Availability details for
Quantity
  • Availability
  • Add to Cart