ENA408615438 Enamine

6-(propan-2-yl)-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD26407730
  • Cas Number : 935542-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136405

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136405
Availability details for
Quantity
  • Availability
  • Add to Cart