ENA408615439 Enamine

4-(3-bromophenyl)-1,2,3,6-tetrahydropyridine hydrochloride

  • MDL number MFCD26407732
  • Cas Number : 92847-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136469
Availability details for
Quantity
  • Availability
  • Add to Cart