ENA408615442 Enamine

1,1,1-trifluoropentan-3-amine hydrochloride

  • MDL number MFCD26407738
  • Cas Number : 101055-07-6
  • Molecular Weight : 177.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 196-200
Iupac Name 1,1,1-trifluoropentan-3-amine hydrochloride
Inchi Code 1S/C5H10F3N.ClH/c1-2-4(9)3-5(6,7)8;/h4H,2-3,9H2,1H3;1H
Inchi Key AQLRLWKAJJIFCS-UHFFFAOYSA-N

Description

Melting Point 196-200
Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-136486

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-136486
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart