ENA408615446 Enamine

4-(butylsulfanyl)-3-(trifluoromethyl)benzoic acid

  • MDL number MFCD26407743
  • Cas Number : 1461705-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136529

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136529
Availability details for
Quantity
  • Availability
  • Add to Cart