ENA408615447 Enamine

methyl 4-(trifluoromethyl)-1,4-dihydroquinoline-3-carboxylate

  • MDL number MFCD26407744
  • Cas Number : 1461705-93-0
  • Molecular Weight : 257.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-(trifluoromethyl)-1,4-dihydroquinoline-3-carboxylate
Inchi Code 1S/C12H10F3NO2/c1-18-11(17)8-6-16-9-5-3-2-4-7(9)10(8)12(13,14)15/h2-6,10,16H,1H3
Inchi Key JHGFTGXVEFPLJN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-136561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136561
Availability details for
Quantity
  • Availability
  • Add to Cart