ENA408615448 Enamine

5-[(4-methylpiperazin-1-yl)methyl]-1,2-oxazole-3-carboxylic acid dihydrochloride

  • MDL number MFCD26407745
  • Cas Number : 1461704-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136572

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136572
Availability details for
Quantity
  • Availability
  • Add to Cart