ENA408615449 Enamine

4-[(3-methyl-1,2,4-oxadiazol-5-yl)methyl]aniline hydrochloride

  • MDL number MFCD26407746
  • Cas Number : 1461709-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136575
Availability details for
Quantity
  • Availability
  • Add to Cart