ENA408615456 Enamine

1-(4-hydrazinylphenyl)ethan-1-one hydrochloride

  • MDL number MFCD26407758
  • Cas Number : 1461713-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136733

Safety Information

signalword Warning
pcodes P201,P202,P261,P264,P270,P271,P280,P281,P301+P312,P302+P352,P304+P340,P305+P351+P338,P308+P313,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-136733
pictograms GHS07,GHS08
hstatements H302,H315,H319,H335,H351
Availability details for
Quantity
  • Availability
  • Add to Cart