ENA408615458 Enamine

1-(cyclopropylmethyl)-6-nitro-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD26407759
  • Cas Number : 1461706-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136779
Availability details for
Quantity
  • Availability
  • Add to Cart