ENA408615461 Enamine

1,4-dimethylpiperidin-4-amine dihydrochloride

  • MDL number MFCD26407763
  • Cas Number : 1461708-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136801
Availability details for
Quantity
  • Availability
  • Add to Cart